2-(Trifluoromethyl)benzophenone 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Phenyl(2-(trifluoromethyl)phenyl)methanone

PC7568-100G PC7568-25G PC7568-5G
76608-648EA 388.12 USD
76608-648 76608-650 76608-652
2-(Trifluoromethyl)benzophenone 98%
2-(Trifluoromethyl)benzophenone
Formula: C₁₄H₉F₃O
MW: 250.22
Melting Pt: 62
Storage Temperature: Ambient
MDL Number: MFCD00000377
CAS Number: 727-99-1
EINECS: 211-972-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR