Sodium chlorodifluoroacetate 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC6460-100G PC6460-25G PC6460-500G
76603-706EA 55.02 USD
76603-706 76603-708 76603-710
Sodium chlorodifluoroacetate 97%
Sodium chlorodifluoroacetate
Formula: C₂ClF₂NaO₂
Melting Pt: 195…198 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064771
CAS Number: 1895-39-2
EINECS: 217-586-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR