Silver hexafluoroantimonate 98%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
Silver hexafluoroantimonate(V)
PC6390-1G
PC6390-25G
PC6390-5G
76618-842EA
22.42
USD
76618-842
76618-844
76618-846
Silver hexafluoroantimonate 98%
Silver hexafluoroantimonate
Lewis acid catalyst for electrophilic halogenation of alkenes, can also promote rearrangements of 3-bromoflavanones to isoflavanones, ask for references
Formula:
AgF₆Sb MW: 343.62 Storage Temperature: Ambient |
MDL Number:
MFCD00003401 CAS Number: 26042-64-8 EINECS: 247-429-1 UN: 3260 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...