3',4',5'-Trifluoropropiophenone 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC7820EA-25G PC7820EA-5G PC7820EA-1G
76618-928EA 347.4 USD
76618-928 76618-930 76618-926
3',4',5'-Trifluoropropiophenone 97%
3',4',5'-Trifluoropropiophenone
Formula: C₉H₇F₃O
MW: 188.15
Storage Temperature: Ambient
CAS Number: 220227-74-7
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR