Trifluoromethanesulfonic acid 99%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Trifluoromethanesulphonic acid, Trifluoromethanesulfonic acid

PC7410-1KG PC7410-250G PC7410-500G
76627-444EA 206.78 USD
76627-444 76627-446 76627-448
Trifluoromethanesulfonic acid 99%
Trifluoromethanesulphonic acid

Useful catalyst /Extremely strong monoprotic acid.

Formula: CHF₃O₃S
MW: 150.08 g/mol
Boiling Pt: 162 °C
Melting Pt: –40 °C
Density: 1.696 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007514
CAS Number: 1493-13-6
EINECS: 216-087-5
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR