2,2,3,3,4,4,4-Heptafluoro-1-butanol 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
1H,1H-Perfluoro-1-butanol, 2,2,3,3,4,4,4-Heptafluorobutan-1-ol, Heptafluoro-1-butanol
PC4390-100G
PC4390-25G
76613-982EA
215.96
USD
76613-982
76613-984
2,2,3,3,4,4,4-Heptafluoro-1-butanol 98%
2,2,3,3,4,4,4-Heptafluoro-1-butanol
Intermediate for speciality coating systems; acrylate and methacrylate esters are available.
Formula:
C₄H₃F₇O MW: 200.06 g/mol Boiling Pt: 96 Density: 1.6 g/cm³ (25 °C) Flash Pt: 25 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004674 CAS Number: 375-01-9 EINECS: 206-782-1 UN: 1987 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...