1,1,1,3,3,3-Hexafluoro-2-propanol 99%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
1,1,1,3,3,3-Hexafluoroisopropyl alcohol, Hexafluoro-2-propanol, Hexafluoroisopropanol, 1,1,1,3,3,3-Hexafluoropropan-2-ol, HFIP
PC4750-1KG
PC4750-250G
PC4750-25G
76623-620EA
190.78
USD
76623-620
76623-622
76623-624
1,1,1,3,3,3-Hexafluoro-2-propanol 99%
1,1,1,3,3,3-Hexafluoro-2-propanol
Versatile intermediate, excellent solvent for polymers, hashigh polarity and low nucleophilicity. Ask for references.
Formula:
C₃H₂F₆O MW: 168.04 g/mol Boiling Pt: 58 °C Melting Pt: –4 °C Density: 1.596 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011651 CAS Number: 920-66-1 EINECS: 213-059-4 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...