α,α,α-Trifluoroacetophenone 99%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2,2,2-Trifluoroacetophenone, Phenacyl trifluoride, Trifluoroacetylbenzene, 1-Phenyl-2,2,2-trifluoro-1-ethanone

PC7200-100G PC7200-25G PC7200-5G
76614-498EA 116.12 USD
76614-498 76614-500 76614-502
α,α,α-Trifluoroacetophenone 99%
α,α,α-Trifluoroacetophenone
Formula: C₈H₅F₃O
MW: 174.12 g/mol
Boiling Pt: 165
Melting Pt: -40
Density: 1.24 g/cm³ (25 °C)
Flash Pt: 41 °C
Storage Temperature: Ambient
MDL Number: MFCD00000420
CAS Number: 434-45-7
EINECS: 207-103-1
UN: 1224
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR