2-Chloro-3-fluoropyridine 99%

Supplier: APOLLO SCIENTIFIC
Danger

PC9542-100G PC9542-25G PC9542-5G
76610-410EA 73.34 USD
76610-410 76610-412 76610-414
2-Chloro-3-fluoropyridine 99%
2-Chloro-3-fluoropyridine
Formula: C₅H₃ClFN
Boiling Pt: 80 °C/80mmHg
Density: 1.323 g/cm³ (25 °C)
Flash Pt: 63 °C
Storage Temperature: Ambient
MDL Number: MFCD03095302
CAS Number: 17282-04-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR