2-Fluorophenol 99%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: o-Fluorophenol

PC4070-100G PC4070-500G
76613-878EA 48.64 USD
76613-878 76613-880
2-Fluorophenol 99%
2-Fluorophenol
Formula: C₆H₅FO
Boiling Pt: 171…172 °C
Melting Pt: 14…16 °C
Density: 1.256 g/cm³ (25 °C)
Flash Pt: 47 °C
Storage Temperature: Ambient
MDL Number: MFCD00002155
CAS Number: 367-12-4
EINECS: 206-681-2
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR