Sanger's reagent 99%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1,3-Dinitro-4-fluorobenzene, 2,4-Dinitrofluorobenzene, 1-Fluoro-2,4-dinitrobenzene

PC3040-100G PC3040-1KG PC3040-500G
76618-136EA 30.56 USD
76618-136 76618-138 76618-140
Sanger's reagent 99%
Sanger's reagent

Nucleophiles can displace the activated fluorine atom.Reagent for labelling of terminal amino acid groups.

Formula: C₆H₃FN₂O₄
MW: 186.1 g/mol
Melting Pt: 28…30 °C
Density: 1.482 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007056
CAS Number: 70-34-8
EINECS: 200-734-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR