Pentafluorobenzoic acid 99%

Supplier: APOLLO SCIENTIFIC
Warning

PC5500-100G PC5500-1KG PC5500-25G
76602-034EA 30.56 USD
76602-034 76602-036 76602-038
Pentafluorobenzoic acid 99%
Pentafluorobenzoic acid
Formula: C₇HF₅O₂
Boiling Pt: 219…221 °C
Melting Pt: 100…104 °C
Density: 1.944 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002406
CAS Number: 602-94-8
EINECS: 210-026-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR