2,3,4,5-Tetrafluorobenzoic acid 99%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
PC6735-100G
PC6735-1KG
PC6735-500G
76603-796EA
26.48
USD
76603-796
76603-798
76603-800
2,3,4,5-Tetrafluorobenzoic acid 99%
2,3,4,5-Tetrafluorobenzoic acid
Formula:
C₇H₂F₄O₂ Melting Pt: 85…87 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009613 CAS Number: 1201-31-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...