4-Ethoxy-3-(trifluoromethyl)benzoyl chloride 97%

Supplier: APOLLO SCIENTIFIC

PC302586-1G PC302586-5G
76613-262EA 54 USD
76613-262 76613-264
4-Ethoxy-3-(trifluoromethyl)benzoyl chloride 97%
4-Ethoxy-3-(trifluoromethyl)benzoyl chloride
Formula: C₁₀H₈ClF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD13196580
CAS Number: 647010-69-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR