2-Fluoro-4-(trifluoromethoxy)benzyl bromide 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC302454-1G PC302454-5G PC302454-10G
76613-248EA 325.39 USD
76613-248 76613-250 76613-246
2-Fluoro-4-(trifluoromethoxy)benzyl bromide 98%
2-Fluoro-4-(trifluoromethoxy)benzyl bromide
Formula: C₈H₅BrF₄O
Storage Temperature: Ambient
MDL Number: MFCD16652442
CAS Number: 1240257-47-9
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR