(4-Chloro-3-(trifluoromethyl)phenyl)acetic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Chloro-3-(trifluoromethyl)phenylacetic acid

PC302061-1G PC302061-5G
76596-712EA 245.14 USD
76596-712 76596-714 76596-716
(4-Chloro-3-(trifluoromethyl)phenyl)acetic acid 97%
(4-Chloro-3-(trifluoromethyl)phenyl)acetic acid
Formula: C₉H₆ClF₃O₂
MW: 238.59
Melting Pt: 82…84 °C
Storage Temperature: Ambient
MDL Number: MFCD04116012
CAS Number: 22902-86-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR