2,4-Difluorophenol 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC2606-100G PC2606-25G
76612-996EA 72.16 USD
76612-996 76612-998
2,4-Difluorophenol 98%
2,4-Difluorophenol
Formula: C₆H₄F₂O
Boiling Pt: 52…53 °C/19mmHg
Melting Pt: 22 °C
Density: 1.362 g/cm³ (25 °C)
Flash Pt: 57 °C
Storage Temperature: Ambient
MDL Number: MFCD00009715
CAS Number: 367-27-1
EINECS: 206-688-0
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR