3-Chloro-α,α,α-trifluorotoluene 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-Chloro-3-(trifluoromethyl)benzene, 3-Chlorobenzotrifluoride

PC1700-100G PC1700-250G
76612-874EA 38.72 USD
76612-874 76612-876
3-Chloro-α,α,α-trifluorotoluene 98%
3-Chloro-α,α,α-trifluorotoluene
Formula: C₇H₄ClF₃
Boiling Pt: 137…138 °C
Melting Pt: –56 °C
Density: 1.331 g/cm³ (25 °C)
Flash Pt: 36 °C
Storage Temperature: Ambient
MDL Number: MFCD00000594
CAS Number: 98-15-7
EINECS: 202-642-9
UN: 2234
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR