1-Bromo-3,4,5-trifluorobenzene 99%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
5-Bromo-1,2,3-trifluorobenzene
PC1575-100G
PC1575-25G
76612-854EA
41.76
USD
76612-854
76612-856
1-Bromo-3,4,5-trifluorobenzene 99%
1-Bromo-3,4,5-trifluorobenzene
Formula:
C₆H₂BrF₃ MW: 210.98 Boiling Pt: 140 Melting Pt: -20 Density: 1.767 g/cm³ (25 °C) Flash Pt: 45 °C Storage Temperature: Ambient |
MDL Number:
MFCD00070738 CAS Number: 138526-69-9 EINECS: 418-480-9 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...