2-Bromo-3'-fluoro-4'-methoxyacetophenone

Supplier: APOLLO SCIENTIFIC
Danger

PC1155-10G PC1155-1G PC1155-5G
76612-768EA 681.48 USD
76612-768 76612-770 76612-772
2-Bromo-3'-fluoro-4'-methoxyacetophenone
2-Bromo-3'-fluoro-4'-methoxyacetophenone
Formula: C₉H₈BrFO₂
Melting Pt: 71…74 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00466240
CAS Number: 350-27-6
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR