Ammonium hexafluorophosphate 99,5%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
PC1102G-100G
PC1102G-25G
PC1102G-500G
76617-690EA
54
USD
76617-690
76617-692
76617-694
Ammonium hexafluorophosphate 99,5%
Ammonium hexafluorophosphate
- Water solubility, 60.4 g/100 g @250C
Formula:
H₄F₆NP Melting Pt: 198 °C (dec.) Density: 2.18 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064642 CAS Number: 16941-11-0 EINECS: 241-009-1 UN: 3260 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...