2-Bromo-2'-chloro-5'-(trifluoromethyl)acetophenone

Supplier: APOLLO SCIENTIFIC
Danger

PC10425-1G PC10425-5G
76612-708EA 216.8 USD
76612-708 76612-710
2-Bromo-2'-chloro-5'-(trifluoromethyl)acetophenone
2-Bromo-2'-chloro-5'-(trifluoromethyl)acetophenone
Formula: C₉H₅BrClF₃O
Melting Pt: 27…29 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03094501
CAS Number: 79779-64-9
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR