N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide (MTBSTFA) 97%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
PC0406-100G
PC0406-25G
PC0406-5G
76612-644EA
227.16
USD
76612-644
76612-646
76612-648
N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide (MTBSTFA) 97%
N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide (MTBSTFA)
GC derivatising agent for silylation of hydroxy, carboxy, thio & primary and secondary amines.
Formula:
C₉H₁₈F₃NOSi Boiling Pt: 168…170 °C Density: 1.023 g/cm³ (25 °C) Flash Pt: 52 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009671 CAS Number: 77377-52-7 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...