3-Fluoro-2-(trifluoromethyl)benzoyl chloride 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC0391-10G PC0391-1G PC0391-5G
76612-638EA 644.82 USD
76612-638 76612-640 76612-642
3-Fluoro-2-(trifluoromethyl)benzoyl chloride 97%
3-Fluoro-2-(trifluoromethyl)benzoyl chloride
Formula: C₈H₃ClF₄O
Storage Temperature: Ambient
MDL Number: MFCD01631458
CAS Number: 261951-82-0
UN: 1760
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR