3-Chloro-2,6-difluorophenol 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC0015-25G PC0015-5G
76622-082EA 337.18 USD
76622-082 76622-084
3-Chloro-2,6-difluorophenol 98%
3-Chloro-2,6-difluorophenol
Formula: C₆H₃ClF₂O
Boiling Pt: 195 °C
Storage Temperature: Ambient
MDL Number: MFCD01631330
CAS Number: 261762-51-0
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR