2-Nitro-5-(1-pyrrolidinyl)aniline

Supplier: APOLLO SCIENTIFIC

OR963708-1G OR963708-5G
76590-670EA 89.64 USD
76590-670 76590-672
2-Nitro-5-(1-pyrrolidinyl)aniline
2-Nitro-5-(1-pyrrolidinyl)aniline
Formula: C₁₀H₁₃N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD00545947
CAS Number: 289913-98-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR