2-Ethoxy-3-pyridineboronic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR9499-1G OR9499-25G OR9499-5G
76590-304EA 20.26 USD
76590-304 76590-308 76590-306
2-Ethoxy-3-pyridineboronic acid
2-Ethoxy-3-pyridineboronic acid
Formula: C₇H₁₀BNO₃
Melting Pt: 102…104 °C
Storage Temperature: Ambient
MDL Number: MFCD06201036
CAS Number: 854373-97-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR