7-Amino-4-methylcoumarin

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 7-Amino-4-methyl-2-benzopyrone, Coumarin 440, Coumarin 120, AMC

OR9400-1G OR9400-25G OR9400-5G
76590-136EA 20.87 USD
76590-136 76590-140 76590-138
7-Amino-4-methylcoumarin
7-Amino-4-methylcoumarin
Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Boiling Pt: 352
Melting Pt: 226
Storage Temperature: Refrigerator
MDL Number: MFCD00006868
CAS Number: 26093-31-2
EINECS: 247-454-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR