2-Bromoterephthalic acid 98+%

Supplier: APOLLO SCIENTIFIC
Warning

OR9323-100G OR9323-25G OR9323-5G
76589-990EA 131.4 USD
76589-990 76589-988 76589-986
2-Bromoterephthalic acid 98+%
2-Bromoterephthalic acid
Formula: C₈H₅BrO₄
Melting Pt: 295…297 °C
Storage Temperature: Ambient
MDL Number: MFCD00002403
CAS Number: 586-35-6
EINECS: 209-572-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR