2-Chlorothiazole 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR9303-25G OR9303-5G
76612-520EA 57.04 USD
76612-520 76612-522
2-Chlorothiazole 98%
2-Chlorothiazole
Formula: C₃H₂ClNS
Boiling Pt: 145…146 °C
Density: 1.393 g/cm³ (25 °C)
Flash Pt: 54.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00210701
CAS Number: 3034-52-4
EINECS: 221-228-9
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR