(2-Hydroxyethyl)hydrazine 92%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
β-Hydroxyethylhydrazine
OR9124-100G
OR9124-10G
OR9124-250G
76612-496EA
58.06
USD
76612-496
76612-498
76612-500
77411-662
(2-Hydroxyethyl)hydrazine 92%
(2-Hydroxyethyl)hydrazine
Formula:
C₂H₈N₂O MW: 76.1 g/mol Boiling Pt: 218…220 °C Melting Pt: –70 °C Density: 1.123 g/cm³ (25 °C) Flash Pt: 77 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007623 CAS Number: 109-84-2 EINECS: 203-711-6 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...