4-tert-Butylbenzylamine 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR9087-25G OR9087-5G
76612-486EA 24.72 USD
76612-486 76612-488 76612-490
4-tert-Butylbenzylamine 98%
4-tert-Butylbenzylamine
Formula: C₁₁H₁₇N
Boiling Pt: 124 °C
Density: 0.927 g/cm³ (25 °C)
Flash Pt: 107 °C
Storage Temperature: Freezer
MDL Number: MFCD00040754
CAS Number: 39895-55-1
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR