5-Hydroxypridine-3-boronic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR908409-100MG OR908409-1G OR908409-250MG
76589-484EA 24.32 USD
76589-484 76589-488 76589-486
5-Hydroxypridine-3-boronic acid 97%
5-Hydroxypridine-3-boronic acid
Formula: C₅H₆BNO₃
Melting Pt: 240…250 °C
Storage Temperature: Refrigerator
MDL Number: MFCD13189132
CAS Number: 1208308-11-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR