4-Aminobenzyl alcohol 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-(Hydroxymethyl)aniline, (4-Aminophenyl)methanol

OR9011-100G OR9011-25G OR9011-5G
76589-312EA 101.88 USD
76589-312 76589-310 76589-308
4-Aminobenzyl alcohol 98%
4-Aminobenzyl alcohol
Formula: C₇H₉NO
MW: 123.15
Melting Pt: 75
Storage Temperature: Refrigerator
MDL Number: MFCD00014782
CAS Number: 623-04-1
EINECS: 210-767-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR