(R)-(+)-2-Pyrrolidinone-5-carboxylic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: D-Pyr-OH, (R)-(+)-2-Pyrrolidone-5-carboxylic acid

OR8966-100G OR8966-25G OR8966-500G
76589-270EA 50.94 USD
76589-270 76589-268 76589-272
(R)-(+)-2-Pyrrolidinone-5-carboxylic acid
(R)-(+)-2-Pyrrolidinone-5-carboxylic acid
Formula: C₅H₇NO₃
Melting Pt: 155…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00066212
CAS Number: 4042-36-8
EINECS: 223-735-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR