2-Chloro-6-methylpyridine

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 6-Chloro-2-picoline

OR8825-100G OR8825-25G OR8825-5G
76589-124EA 80.48 USD
76589-124 76589-122 76589-120
2-Chloro-6-methylpyridine
2-Chloro-6-methylpyridine
Formula: C₆H₆ClN
MW: 127.57 g/mol
Boiling Pt: 64
Density: 1.167 g/cm³ (25 °C)
Flash Pt: 74 °C
Storage Temperature: Ambient
MDL Number: MFCD00006245
CAS Number: 18368-63-3
EINECS: 242-241-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR