3-Formylphenylboronic acid

Supplier: APOLLO SCIENTIFIC
Danger

OR6998-100G OR6998-25G
76621-842EA 82.52 USD
76621-842 76621-844
3-Formylphenylboronic acid
3-Formylphenylboronic acid
Formula: C₇H₇BO₃
Melting Pt: 109…113 °C
Storage Temperature: Ambient
MDL Number: MFCD00161356
CAS Number: 87199-16-4
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR