Cyclopropylméthanol 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Cyclopropyl carbinol, Cyclopropanemethanol

OR6507-100G OR6507-25G OR6507-500G
76612-282EA 68.24 USD
76612-282 76612-284 76612-286
Cyclopropylméthanol 98%
Cyclopropylméthanol
Formula: C₄H₈O
Boiling Pt: 123…124 °C
Melting Pt: –60 °C
Density: 0.89 g/cm³ (25 °C)
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00001309
CAS Number: 2516-33-8
UN: 2920
ADR: 8+3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR