2-Bromo-3-nitrobenzoic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR6235-100G OR6235-25G OR6235-5G
76594-572EA 186.42 USD
76594-572 76594-570 76594-568
2-Bromo-3-nitrobenzoic acid 98%
2-Bromo-3-nitrobenzoic acid
Formula: C₇H₄BrNO₄
MW: 246.02 g/mol
Melting Pt: 184…188 °C
Storage Temperature: Ambient
MDL Number: MFCD00074899
CAS Number: 573-54-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR