Hydroxylammonium chloride 99%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
Hydroxylamine HCl, Hydroxylamine hydrochloride
OR62004-1KG
OR62004-5KG
76617-180EA
69.26
USD
76617-180
76617-182
Hydroxylammonium chloride 99%
Hydroxylammonium chloride
Formula:
NH₂OH·HCl MW: 69.49 g/mol Boiling Pt: 306 Melting Pt: 151 (dec.) Density: 1.67 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00051089 CAS Number: 5470-11-1 UN: 2923 ADR: 8+6.1 III Merck Index: 13,04853 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...