3-Octanone

Supplier: APOLLO SCIENTIFIC
Warning

OR6025-100G OR6025-25G
76612-196EA 33.62 USD
76612-196 76612-198
3-Octanone
3-Octanone
Formula: C₈H₁₆O
Boiling Pt: 167…168 °C
Density: 0.822 g/cm³ (25 °C)
Flash Pt: 46 °C
Storage Temperature: Ambient
MDL Number: MFCD00009515
CAS Number: 106-68-3
UN: 2271
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR