7-Bromo-1-heptene 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR60221-100G OR60221-25G OR60221-5G
76612-190EA 156.88 USD
76612-190 76612-192 76612-194
7-Bromo-1-heptene 98%
7-Bromo-1-heptene
Formula: C₇H₁₃Br
Boiling Pt: 27 °C/1mmHg
Density: 1.162 g/cm³ (25 °C)
Flash Pt: 58.3 °C
Storage Temperature: Ambient
MDL Number: MFCD01631137
CAS Number: 4117-09-3
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR