7-Amino-2-methylindazole

Supplier: APOLLO SCIENTIFIC
Warning

OR60014-250MG OR60014-5G OR60014-1G
76593-596EA 20.26 USD
76593-596 76593-600 76593-598
7-Amino-2-methylindazole
7-Amino-2-methylindazole
Formula: C₈H₉N₃
MW: 147.18 g/mol
Melting Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD03787880
CAS Number: 90223-02-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR