1-(p-Toluenesulfonyl)imidazole
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
1-(P-Toluenesulfonyl)imidazole, 1-(P-Toluenesulphonyl)imidazole, 1-Tosylimidazole
OR59904-100G
OR59904-25G
76593-516EA
54
USD
76593-516
76593-514
76593-512
1-(p-Toluenesulfonyl)imidazole
1-(p-Toluenesulphonyl)imidazole
Formula:
C₁₀H₁₀N₂O₂S MW: 222.26 g/mol Melting Pt: 76…78 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005285 CAS Number: 2232-08-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...