Potassium xanthogenate (potassium-O-ethyl dithiocarbonate)

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Potassium-O-ethyldithiocarbonate, Potassium ethyl xanthate, Ethylxanthic acid potassium salt

OR59885-100G OR59885-500G
76625-372EA 109.52 USD
76625-372 76625-374 76625-376
Potassium xanthogenate (potassium-O-ethyl dithiocarbonate)
Potassium xanthogenate (Potassium O-ethyl dithiocarbonate)
Formula: C₃H₅KOS₂
Melting Pt: 210 °C
Density: 1.558 g/cm³ (25 °C)
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00004931
CAS Number: 140-89-6
UN: 3342
ADR: 4.2,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR