Indole-3-carboxylic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1H-Indole-3-carboxylic acid

OR5873-100G OR5873-25G OR5873-500G
76593-166EA 35.66 USD
76593-166 76593-164 76593-168
Indole-3-carboxylic acid 98%
Indole-3-carboxylic acid
Formula: C₉H₇NO₂
Melting Pt: 232…239 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00005624
CAS Number: 771-50-6
EINECS: 212-231-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR