Benzoylacetonitrile 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR5834-100G OR5834-250G OR5834-25G
76593-108EA 124.28 USD
76593-108 76593-110 76593-106
Benzoylacetonitrile 95%
Benzoylacetonitrile
Formula: (C₆H₅)C(O)CH₂CN
Boiling Pt: 159…160 °C/10mmHg
Melting Pt: 82…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00001942
CAS Number: 614-16-4
EINECS: 210-369-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR