2-Amino-6-chloro-3-nitropyridine 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 6-Chloro-3-nitro-2-pyridylamine, 6-Chloro-3-nitropyridin-2-amine

OR5822-100G OR5822-25G OR5822-5G
76593-078EA 51 USD
76593-078 76593-076 76593-074
2-Amino-6-chloro-3-nitropyridine 98%
2-Amino-6-chloro-3-nitropyridine
Formula: C₅H₄ClN₃O₂
MW: 173.56
Melting Pt: 194…196 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00044350
CAS Number: 27048-04-0
EINECS: 248-188-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR