2-Chlorobenzyl alcohol 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (2-Chlorophenyl)methanol

OR5737-100G OR5737-25G
76592-980EA 23.38 USD
76592-980 76592-978
2-Chlorobenzyl alcohol 98%
2-Chlorobenzyl alcohol
Formula: C₇H₇ClO
MW: 142.58 g/mol
Boiling Pt: 227
Melting Pt: 69…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00004604
CAS Number: 17849-38-6
EINECS: 241-801-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR