1-(2-Methoxyphenyl)-2-thiourea

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: N-(2-Methoxyphenyl)thiourea, 1-(2-Methoxyphenyl)thiourea, 2-Methoxyphenylthiourea

OR5634-1G OR5634-5G
76617-032EA 20.87 USD
76617-032 76617-034
1-(2-Methoxyphenyl)-2-thiourea
1-(2-Methoxyphenyl)-2-thiourea
Formula: (C₆H₄(OCH₃))NC(S)NH₂
Storage Temperature: Ambient
MDL Number: MFCD00022162
CAS Number: 1516-37-6
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR